missing translation for 'onlineSavingsMsg'
Learn More

Butyl Gallate 98.0+%, TCI America™

Catalog No. G018625G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
G018625G 25 g
1 options

Catalog No. G018625G

Supplier: TCI America G018625G

Only null left

Chemical Identifiers

CAS 1083-41-6
Molecular Formula C11H14O5
Molecular Weight (g/mol) 226.228
MDL Number MFCD00016432
InChI Key XOPOEBVTQYAOSV-UHFFFAOYSA-N
Synonym Gallic Acid Butyl Ester
PubChem CID 14128
IUPAC Name butyl 3,4,5-trihydroxybenzoate
SMILES CCCCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 144°C
Color White
Quantity 25 g
Formula Weight 226.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Butyl Gallate
EINECSNumber (3)-1583
RTECSNumber LW7600000
TSCA No