missing translation for 'onlineSavingsMsg'
Learn More

Butyl Formate 95.0+%, TCI America™

$55.92 - $378.40

Chemical Identifiers

CAS 592-84-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00003296
InChI Key NMJJFJNHVMGPGM-UHFFFAOYSA-N
Synonym n-butyl formate, formic acid, butyl ester, butyl methanoate, n-butyl methanoate, formic acid butyl ester, n-butylformate, butylester kyseliny mravenci, fema no. 2196, unii-0y9mz7vm9p, butylester kyseliny mravenci czech
PubChem CID 11614
IUPAC Name butyl formate
SMILES CCCCOC=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F005125ML
View Documents
TCI America
F005125ML
25 mL N/A
Only null left
 
F0051500ML
View Documents
TCI America
F0051500ML
500 mL N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

592-84-7
102.13
NMJJFJNHVMGPGM-UHFFFAOYSA-N
11614
CCCCOC=O
C5H10O2
MFCD00003296
n-butyl formate, formic acid, butyl ester, butyl methanoate, n-butyl methanoate, formic acid butyl ester, n-butylformate, butylester kyseliny mravenci, fema no. 2196, unii-0y9mz7vm9p, butylester kyseliny mravenci czech
butyl formate

Specifications

592-84-7
Yellow
C5H10O2
25 mL
n-butyl formate, formic acid, butyl ester, butyl methanoate, n-butyl methanoate, formic acid butyl ester, n-butylformate, butylester kyseliny mravenci, fema no. 2196, unii-0y9mz7vm9p, butylester kyseliny mravenci czech
CCCCOC=O
102.13
102.13
Liquid
-91°C
107°C
MFCD00003296
1128
NMJJFJNHVMGPGM-UHFFFAOYSA-N
butyl formate
11614
≥95.0% (GC)
Butyl Formate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0684

RTECSNumber : LQ5500000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'