missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Cyanoacetate 98.0+%, TCI America™
$41.02 - $473.48
Chemical Identifiers
| CAS | 5459-58-5 |
|---|---|
| Molecular Formula | C7H11NO2 |
| Molecular Weight (g/mol) | 141.17 |
| MDL Number | MFCD00001941 |
| InChI Key | DJACTCNGCHPGOI-UHFFFAOYSA-N |
| Synonym | butyl cyanoacetate, n-butyl cyanoacetate, acetic acid, cyano-, butyl ester, unii-f9h6l20voh, cyanoacetic acid butyl ester, cyanoacetic acid n-butyl ester, acetic acid, 2-cyano-, butyl ester, f9h6l20voh, butyl 2-cyanoethanoate, acmc-209lho |
| PubChem CID | 79571 |
| IUPAC Name | butyl 2-cyanoacetate |
| SMILES | CCCCOC(=O)CC#N |
Chemical Identifiers
| 5459-58-5 | |
| 141.17 | |
| DJACTCNGCHPGOI-UHFFFAOYSA-N | |
| 79571 | |
| CCCCOC(=O)CC#N |
| C7H11NO2 | |
| MFCD00001941 | |
| butyl cyanoacetate, n-butyl cyanoacetate, acetic acid, cyano-, butyl ester, unii-f9h6l20voh, cyanoacetic acid butyl ester, cyanoacetic acid n-butyl ester, acetic acid, 2-cyano-, butyl ester, f9h6l20voh, butyl 2-cyanoethanoate, acmc-209lho | |
| butyl 2-cyanoacetate |
Specifications
| 5459-58-5 | |
| 231°C | |
| MFCD00001941 | |
| butyl cyanoacetate, n-butyl cyanoacetate, acetic acid, cyano-, butyl ester, unii-f9h6l20voh, cyanoacetic acid butyl ester, cyanoacetic acid n-butyl ester, acetic acid, 2-cyano-, butyl ester, f9h6l20voh, butyl 2-cyanoethanoate, acmc-209lho | |
| CCCCOC(=O)CC#N | |
| 141.17 | |
| 141.17 | |
| Liquid |
| Yellow | |
| C7H11NO2 | |
| 25 g | |
| DJACTCNGCHPGOI-UHFFFAOYSA-N | |
| butyl 2-cyanoacetate | |
| 79571 | |
| ≥98.0% (GC) | |
| Butyl Cyanoacetate |
Safety and Handling
EINECSNumber : (2)-1549
RTECSNumber : AG3692500
TSCA : Yes