missing translation for 'onlineSavingsMsg'
Learn More

Butyl Crotonate 97.0+%, TCI America™

Catalog No. C041725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C041725ML 25 mL
C0417500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C041725ML Supplier TCI America Supplier No. C041725ML

Chemical Identifiers

CAS 7299-91-4
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00053794
InChI Key GWCTUKHUBWMAMI-GQCTYLIASA-N
Synonym Crotonic Acid Butyl Ester
PubChem CID 5366039
IUPAC Name butyl (2E)-but-2-enoate
SMILES CCCCOC(=O)\C=C\C

Specifications

Color Colorless
Boiling Point 180°C
Quantity 25 mL
Formula Weight 142.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Crotonate
EINECSNumber (2)-0966
RTECSNumber GQ3200000
TSCA No