Learn More
Chemical Identifiers
| CAS | 592-34-7 |
|---|---|
| Molecular Formula | C5H9ClO2 |
| Molecular Weight (g/mol) | 136.58 |
| MDL Number | MFCD00000650 |
| InChI Key | NRDQFWXVTPZZAZ-UHFFFAOYSA-N |
| Synonym | butyl chloroformate, n-butyl chloroformate, butylchloroformate, carbonochloridic acid, butyl ester, chloroformic acid butyl ester, butyl chlorocarbonate, n-butyl chlorocarbonate, formic acid, chloro-, butyl ester, butyl-chloroformate, butylchloro-formate |
| PubChem CID | 61140 |
| IUPAC Name | butyl carbonochloridate |
| SMILES | CCCCOC(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC180170250
|
N/A | 25mL | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC180170010
|
N/A | 1L | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Coupling reagentChemical Identifiers
| C5H9ClO2 | |
| MFCD00000650 | |
| butyl chloroformate, n-butyl chloroformate, butylchloroformate, carbonochloridic acid, butyl ester, chloroformic acid butyl ester, butyl chlorocarbonate, n-butyl chlorocarbonate, formic acid, chloro-, butyl ester, butyl-chloroformate, butylchloro-formate | |
| butyl carbonochloridate |
Specifications
| 592-34-7 | |
| 142.0°C | |
| Authentic | |
| C5H9ClO2 | |
| ClCO2(CH2)3CH3 | |
| 03, II, 11 | |
| butyl chloroformate, n-butyl chloroformate, butylchloroformate, carbonochloridic acid, butyl ester, chloroformic acid butyl ester, butyl chlorocarbonate, n-butyl chlorocarbonate, formic acid, chloro-, butyl ester, butyl-chloroformate, butylchloro-formate | |
| NRDQFWXVTPZZAZ-UHFFFAOYSA-N | |
| butyl carbonochloridate | |
| 61140 | |
| 136.58 | |
| Butyl chloroformate, 98% |
| 1.0600g/mL | |
| 39°C | |
| 97.5% min. (GC) | |
| 1.4110 to 1.4130 | |
| MFCD00000650 | |
| 1.06 | |
| Solubility in water: decomposes in water. Other solubilities: soluble in butanol,ether,benzol,soluble in most common organic solvents | |
| CCCCOC(=O)Cl | |
| 136.58 | |
| 0.95 mPa.s (20°C) | |
| 98% |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-750-5
TSCA : TSCA
RUO – Research Use Only