missing translation for 'onlineSavingsMsg'
Learn More

Butyl Butyrate 99.0+%, TCI America™

Catalog No. B075725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B075725ML 25 mL
B0757500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B075725ML Supplier TCI America Supplier No. B075725ML

Chemical Identifiers

CAS 109-21-7
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00009450
InChI Key XUPYJHCZDLZNFP-UHFFFAOYSA-N
Synonym butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate
PubChem CID 7983
ChEBI CHEBI:87429
IUPAC Name butyl butanoate
SMILES CCCCOC(=O)CCC

Specifications

Melting Point -92°C
Color Colorless
Boiling Point 164°C
Quantity 25 mL
UN Number 3272
Formula Weight 144.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Butyrate
EINECSNumber (2)-0779
RTECSNumber ES8120000
TSCA Yes