missing translation for 'onlineSavingsMsg'
Learn More

Butyl Butyrate, 98%, Spectrum™ Chemical

Catalog No. 18609378
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609378 200 L Drum
18609380 500 mL Amber Glass Bottle
18609379 4 L Amber Glass Bottle
18609377 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609378 Supplier Spectrum Chemical Mfg Cor Supplier No. B1183200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-21-7

Specifications

Acidity 1
CAS Max % 1
Packaging Drum
Refractive Index 1.405-1.407
Quantity 200 L
Specific Gravity 0.867 - 0.871
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.