missing translation for 'onlineSavingsMsg'
Learn More

Butyl Alcohol, Reagent, ACS, 99.4%, Spectrum™ Chemical

Catalog No. 18601153
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601153 100 mL Amber Glass Bottle
18601154 500 mL Amber Glass Bottle
18601156 1 L Amber Glass Bottle
18601155 6 x 500 mL Amber Glass Bottle
18601157 4 L Amber Glass Bottle
18601158 6 x 1 L Amber Glass Bottle
18601159 4 x 4 L Amber Glass Bottle
18601160 20 L Steel Can
18601161 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18601153 Supplier Spectrum Chemical Mfg Cor Supplier No. B1185100ML

B1185, 71-36-3, C4H10O

Spectrum™ Chemical Butyl Alcohol, Reagent, ACS is more commonly known as butanol. It is also widely used as a solvent in textile and chemical fabrication processes. It can be used as a paint thinner and base for perfumes. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

CAS Max % 100%
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 0.005%
Titratable Acid 0.0008 meq/g
Water 0.1%
UN Number UN1120
Boiling Point 117°C
Percent Purity 99.4%
Grade Reagent
Chemical Name or Material Butyl Alcohol
Show More Show Less