missing translation for 'onlineSavingsMsg'
Learn More

Butyl Alcohol ACS, MilliporeSigma™

Catalog No. MBX17806
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MBX17806 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MBX17806 Supplier Technidata Supplier No. BX17806
Only null left

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Quantity 500 mL
Packaging Glass Bottle
Linear Formula CH3(CH2)2CH2OH
Formula Weight 74.12
Grade ACS
Chemical Name or Material Butyl Alcohol