missing translation for 'onlineSavingsMsg'
Learn More

Butyl Alcohol, 99.7% min, OmniSolv, MilliporeSigma™

Catalog No. 361020556
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361020556 1 L
MBX17771 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361020556 Supplier Technidata Supplier No. BX17776

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Density 0.81g/cm3 (20°C)
Quantity 1 L
Explosion Limit 1.4 to 11.3%(V)
Ignition Point 340°C
Physical Form Liquid
Assay Percent Range 99.7% min.
Vapor Pressure 6.7 hPa (20°C)
Linear Formula CH3(CH2)3OH
Formula Weight 74.12
Chemical Name or Material Butyl Alcohol
Recommended Storage Store without restrictions