missing translation for 'onlineSavingsMsg'
Learn More

Butyl Acrylate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. p-7116735
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
A014225ML 25 mL
A0142500ML 500 mL
2 options

Catalog No. A014225ML

Supplier: TCI America A014225ML

Only null left

Chemical Identifiers

CAS 141-32-2
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00009446
InChI Key CQEYYJKEWSMYFG-UHFFFAOYSA-N
Synonym butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove
PubChem CID 8846
ChEBI CHEBI:3245
IUPAC Name butyl prop-2-enoate
SMILES CCCCOC(=O)C=C

Specifications

Melting Point -64°C
Color Colorless
Boiling Point 145°C
Quantity 25 mL
UN Number 2348
Formula Weight 128.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-0989
RTECSNumber UD3150000
TSCA Yes