missing translation for 'onlineSavingsMsg'
Learn More

Butyl Acrylate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. A014225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A014225ML 25 mL
A0142500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A014225ML Supplier TCI America Supplier No. A014225ML

Chemical Identifiers

CAS 141-32-2
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00009446
InChI Key CQEYYJKEWSMYFG-UHFFFAOYSA-N
Synonym butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove
PubChem CID 8846
ChEBI CHEBI:3245
IUPAC Name butyl prop-2-enoate
SMILES CCCCOC(=O)C=C

Specifications

Melting Point -64°C
Color Colorless
Boiling Point 145°C
Quantity 25 mL
UN Number 2348
Formula Weight 128.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-0989
RTECSNumber UD3150000
TSCA Yes