missing translation for 'onlineSavingsMsg'
Learn More

Acétate de butyle 99,0+%, TCI America™

Numéro de catalogue. A002425ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A002425ML 25 mL
A0024500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A002425ML Fournisseur TCI America Code fournisseur A002425ML

Identifiants chimiques

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name Acétate de butyle
SMILES CCCCOC(C)=O

Spécifications

Melting Point -78°C
Color Incolore
Boiling Point 126°C
Quantity 25 mL
UN Number 1123
Formula Weight 116.16
Percent Purity ≥99.0% (GC)
Physical Form Liquide
Chemical Name or Material Butyl Acetate
EINECSNumber (2)-0731
RTECSNumber AF7350000
TSCA Yes