missing translation for 'onlineSavingsMsg'
Learn More

Butyl Acetate, puriss. p.a., ACS Reagent, ≥99.5% (GC), Honeywell Riedel-de Haën™

Catalog No. p-7120267
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046924 1 L Glass Bottle
60046925 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046924 Supplier Honeywell Chemicals Supplier No. 458601L
Only null left

Puriss. p.a., ACS Reagent

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -78°C
Density 0.88g/mL at 25°C
Boiling Point 126°C
Explosion Limit 0.076
Flash Point 23°C (73.4°F)
Ignition Point 790°F
Packaging Glass Bottle
Refractive Index n20/D 1.394
Linear Formula CH3COO(CH2)3CH3
Quantity 1 L
UN Number UN1123
Residue after Evaporation 0 to 0.001%
Beilstein 1741921
Vapor Density 4 (vs air)
Vapor Pressure 15mmHg (25°C), 8mmHg (20°C)
Formula Weight 116.16g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material Butyl acetate
Show More Show Less
Hazard Statement P210-P370 + P378
DOTInformation class 3 - PG 3 - Butyl acetates
RTECSNumber 204-658-1