missing translation for 'onlineSavingsMsg'
Learn More

Butyl 2-Chloroethyl Ether 98.0+%, TCI America™

Catalog No. p-7124835
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11715ML 5 mL
C117125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11715ML Supplier TCI America Supplier No. C11715ML
Only null left

Chemical Identifiers

CAS 10503-96-5
Molecular Formula C6H13ClO
Molecular Weight (g/mol) 136.62
MDL Number MFCD00059051
InChI Key OJMVRGXLTANFRG-UHFFFAOYSA-N
Synonym 2-Chloroethyl Butyl Ether
PubChem CID 545871
IUPAC Name 1-(2-chloroethoxy)butane
SMILES CCCCOCCCl

Specifications

Color Yellow
Boiling Point 50°C
Quantity 5 mL
UN Number 3271
Formula Weight 136.62
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl 2-Chloroethyl Ether