missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Butterfly-Style Shoe Cover Universal

Catalog No. 19165003
Change view
Click to view available options
Size:
Universal
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19165003 Universal
19165003XL X-Large
191650032X 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19165003 Supplier Fisherbrand™ Supplier No. SHT1912B
Only null left

Fluid resistant material provides slip and fall protection in both wet and dry conditions.

  • Anti-skid
  • Fluid resistant
  • Performs well in low-level standing water
  • Butterfly style with sonic welded seams

Specifications

Size Universal
Material Polyethylene, Polypropylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Anti-skid
Seam Type Ultrasonically Welded
For Use With (Application) Controlled and Critical Environments
Packaging Quantity 200/Cs.
Length (English) 15.5 in.
Height (English) 6.5 in.
Resistance Fluid
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 100 (ISO 5)
Show More Show Less