missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butoxyacetic Acid 98.0+%, TCI America™
$72.70 - $257.24
Chemical Identifiers
| CAS | 2516-93-0 |
|---|---|
| Molecular Formula | C6H12O3 |
| Molecular Weight (g/mol) | 132.159 |
| MDL Number | MFCD00067044 |
| InChI Key | AJQOASGWDCBKCJ-UHFFFAOYSA-N |
| Synonym | butoxyacetic acid, n-butoxyacetic acid, acetic acid, butoxy, ccris 7195, acetic acid, 2-butoxy, 2-n-butoxy acetic acid, butoxyaceticacid, 3-oxaheptanoic acid, acmc-1cb1h, ajqoasgwdcbkcj-uhfffaoysa |
| PubChem CID | 41958 |
| IUPAC Name | 2-butoxyacetic acid |
| SMILES | CCCCOCC(=O)O |
Chemical Identifiers
| 2516-93-0 | |
| 132.159 | |
| AJQOASGWDCBKCJ-UHFFFAOYSA-N | |
| 41958 | |
| CCCCOCC(=O)O |
| C6H12O3 | |
| MFCD00067044 | |
| butoxyacetic acid, n-butoxyacetic acid, acetic acid, butoxy, ccris 7195, acetic acid, 2-butoxy, 2-n-butoxy acetic acid, butoxyaceticacid, 3-oxaheptanoic acid, acmc-1cb1h, ajqoasgwdcbkcj-uhfffaoysa | |
| 2-butoxyacetic acid |
Specifications
| 2516-93-0 | |
| 234°C | |
| MFCD00067044 | |
| 3265 | |
| AJQOASGWDCBKCJ-UHFFFAOYSA-N | |
| 2-butoxyacetic acid | |
| 41958 | |
| ≥98.0% (GC,T) | |
| Butoxyacetic Acid |
| Colorless | |
| C6H12O3 | |
| 1 g | |
| butoxyacetic acid, n-butoxyacetic acid, acetic acid, butoxy, ccris 7195, acetic acid, 2-butoxy, 2-n-butoxy acetic acid, butoxyaceticacid, 3-oxaheptanoic acid, acmc-1cb1h, ajqoasgwdcbkcj-uhfffaoysa | |
| CCCCOCC(=O)O | |
| 132.159 | |
| 132.16 | |
| Liquid |
Safety and Handling
TSCA : No