missing translation for 'onlineSavingsMsg'
Learn More

Butoxyacetic Acid 98.0+%, TCI America™

Catalog No. B14675G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14675G 5 g
B14671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14675G Supplier TCI America Supplier No. B14675G
Only null left

Chemical Identifiers

CAS 2516-93-0
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00067044
InChI Key AJQOASGWDCBKCJ-UHFFFAOYSA-N
Synonym butoxyacetic acid, n-butoxyacetic acid, acetic acid, butoxy, ccris 7195, acetic acid, 2-butoxy, 2-n-butoxy acetic acid, butoxyaceticacid, 3-oxaheptanoic acid, acmc-1cb1h, ajqoasgwdcbkcj-uhfffaoysa
PubChem CID 41958
IUPAC Name 2-butoxyacetic acid
SMILES CCCCOCC(=O)O

Specifications

Color Colorless
Boiling Point 234°C
Quantity 5 g
UN Number 3265
Formula Weight 132.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Butoxyacetic Acid