missing translation for 'onlineSavingsMsg'
Learn More

Buspirone hydrochloride

Catalog No. p-7041850
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6047603 1 g
AAJ6047606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6047603 Supplier Thermo Scientific Chemicals Supplier No. J6047603

CAS: 33386-08-2 | C21H32ClN5O2 | 421.97 g/mol

It is widely used in cell signaling and neuroscience research. Buspirone has been used to mitigate the electrically stimulated 5-hydroxytryptamine outflow from rat cortical slices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is widely used in cell signaling and neuroscience research. Buspirone has been used to mitigate the electrically stimulated 5-hydroxytryptamine outflow from rat cortical slices.

Solubility
Soluble in methanol (50 mg/ml), water (partly), DMSO (100 mM).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 33386-08-2
Molecular Formula C21H32ClN5O2
Molecular Weight (g/mol) 421.97
MDL Number MFCD00078569
InChI Key RICLFGYGYQXUFH-UHFFFAOYSA-N
Synonym buspirone hydrochloride, buspar, buspirone hcl, narol, buspimen, ansitec, anxinil, anxiolan, busirone, buspinol
PubChem CID 36431
ChEBI CHEBI:3224
IUPAC Name 8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
SMILES C1CCC2(C1)CC(=O)N(C(=O)C2)CCCCN3CCN(CC3)C4=NC=CC=N4.Cl

Specifications

Color White
Quantity 1 g
UN Number UN2811
Merck Index 14,1504
Solubility Information Soluble in methanol (50mg/ml),water (partly),DMSO (100 mM).
Formula Weight 421.96
Physical Form Powder
Chemical Name or Material Buspirone hydrochloride
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 251-489-4
RTECSNumber CL9915000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only