Learn More
Fine Science Tools Burrs for Micro Drill, Tungsten Carbide, 0.5 mm Tip Dia, 44 mm, 5/pk
Catalog No. NC1744244
Encompass
Tip Diameter: 0.5mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Tungsten Carbide/ Length: 44mm/ Autoclave Safe: Yes
Catalog No. NC1744244 Supplier Fine Science Tools Supplier No. 1900905
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tip Diameter: 0.5mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Tungsten Carbide/ Length: 44mm/ Autoclave Safe: Yes