missing translation for 'onlineSavingsMsg'
Learn More

Burrell Scientific Wrist Action™ Laboratory Shaker Accessory, Volumetric SUREGRIP™ Clamp

For use with Wrist Action Laboratory Shaker

Supplier:  Burrell Scientific 0757771800

Catalog No. 14-400-144


Only null left

Description

Description

  • Maximum adjustability and security
  • Holds vessels up to 500mL
  • Perfect for Volumetric Flasks
  • Designed with bottom support to stabilize larger vessels
  • Fully adjustable
  • Material: Aluminum
  • Color: Silver and red
Specifications

Specifications

Clamp
Wrist Action Laboratory Shaker
2
1 Year
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions