missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Burette Holder

Catalog No. 01257568
Change view
Click to view available options
Type: Burette Holder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01257568 Type: Burette Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01257568 Supplier Fisherbrand™ Supplier No. CB17030

Burette Holder is a spring loaded accessory for laboratory

  • Clamps are die-cast from aluminum and have rubber grips to prevent slippage
  • Loaded springs create enough pressure to hold burettes securely
  • Powder coated finish on surface prevents from scratches and ensures long time use
  • Do not obscure the buret graduation lines and numbers

Specifications

Color Silver
Length (Metric) 250 mm
For Use With (Equipment) Burettes
No. per Case 50
Length (English) 10 in.
Type Buret Holder