missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Burette Clamp

Catalog No. S23794
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S23794 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23794 Supplier Eisco™ Supplier No. CH0258A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

With high strength alloy jaws.

  • Suitable for all burettes up to 100mL
  • With high strength alloy jaws

Specifications

Type Burette Clamp
Material Alloy
For Use With (Equipment) Burettes up to 100mL
Color Blue
Jaw Type High-strength Alloy