missing translation for 'onlineSavingsMsg'
Learn More

Bupropion Hydrochloride 98.0+%, TCI America™

Catalog No. B36495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36495G 5 g
B36491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36495G Supplier TCI America Supplier No. B36495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31677-93-7
Molecular Formula C13H19Cl2NO
Molecular Weight (g/mol) 276.201
MDL Number MFCD00055209
InChI Key HEYVINCGKDONRU-UHFFFAOYSA-N
Synonym bupropion hydrochloride, wellbutrin, 2-tert-butylamino-1-3-chlorophenyl propan-1-one hydrochloride, bupropion hcl, zyban, wellbutrin xl, bupropion hydrocloride, wellbutrin sr, budeprion, forfivo xl
PubChem CID 62884
IUPAC Name 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride
SMILES CC(C(=O)C1=CC(=CC=C1)Cl)NC(C)(C)C.Cl

Specifications

Melting Point 222°C
Color White
Quantity 5 g
Formula Weight 276.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bupropion Hydrochloride
RTECSNumber UG8858000
TSCA No