missing translation for 'onlineSavingsMsg'
Learn More

Bupropion hydrochloride, 99%

Catalog No. aaj6110503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6110503 1 g
AAJ61105MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6110503 Supplier Thermo Scientific Chemicals Supplier No. J6110503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A selective inhibitor of dopamine uptake | CAS: 31677-93-7 | C13H19Cl2NO | 276.201 g/mol

Bupropion HC is used as an inhibitor of dopamine and SLC6A2. Bupropion is efficacious as an antidepressant because of its effects on nicotinic receptors, it is also used to promote smoking cessation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bupropion HC is used as an inhibitor of dopamine and SLC6A2. Bupropion is efficacious as an antidepressant because of its effects on nicotinic receptors, it is also used to promote smoking cessation.

Solubility
Soluble in water (>25 mg/ml), ethanol 193mg/ml, 0.1 N HCl 333 mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 31677-93-7
Molecular Formula C13H19Cl2NO
Molecular Weight (g/mol) 276.201
MDL Number MFCD00055209
InChI Key HEYVINCGKDONRU-UHFFFAOYSA-N
Synonym bupropion hydrochloride, wellbutrin, 2-tert-butylamino-1-3-chlorophenyl propan-1-one hydrochloride, bupropion hcl, zyban, wellbutrin xl, bupropion hydrocloride, wellbutrin sr, budeprion, forfivo xl
PubChem CID 62884
IUPAC Name 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride
SMILES CC(C(=O)C1=CC(=CC=C1)Cl)NC(C)(C)C.Cl

Specifications

Melting Point 233°C to 234°C
Color White
Quantity 1 g
Merck Index 14,1499
Solubility Information Soluble in water (>25mg/ml),ethanol 193mg/ml,0.1 N HCl 333mg/ml
Formula Weight 276.2
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Bupropion hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 250-759-9
RTECSNumber UG8858000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only