missing translation for 'onlineSavingsMsg'
Learn More

Buprenorphine hydrochloride Solution, MilliporeSigma™ Supelco™

Catalog No. 111016606
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016606 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016606 Supplier Merck Emd Millipore Supplier No. B75361ML
Only null left
Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 53152-21-9
Flash Point 11°C
Linear Formula C29H41 NO4 · HCl
MDL Number MFCD00084836
Type Analytical Standard
Industry Type Drug Analysis
Quantity 1 mL
Formula Weight 504.10
Grade Analytical Standard