missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Bumper Guard For Volumetric Flask

Catalog No. S16070
Change view
Click to view available options
Capacity (Metric) Media:
25 mL
100 mL
250 mL
500 mL
1000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Media
S16070 250 mL
S15908 100 mL
S15965 500 mL
S15741 1000 mL
S15886 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S16070 Supplier United Scientific™ Supplier No. BMP250PK5
Only null left

These guards help prevent volumetric glassware from breaking when tipping over.

  • Silicone bumpers slip easily onto any volumetric flask. In the event of a flask being accidentally tipped over, the wide bumpers cushion the neck from hitting the table surface.
  • Color coded and available in five different sizes.
  • Wide design to cushion flasks if tipped over
  • Made of high-quality silicone

Specifications

Capacity (Metric) Media 250 mL
For Use With (Equipment) Volumetric Flask
Product Type Bumper Guard