missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Nomex™ IIIA HRC1 Lab Coats

Catalog No. 191661070
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
191661070 Large
191661068 Small
191661072 2X-Large
191661071 X-Large
19295002 3X-Large
191661069 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 191661070 Supplier Bulwark™ Supplier No. KNL2RB RG L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Constructed with flame-resistant Nomex IIIA fabric.

  • Arc Rating: ATPV 5.6 calories/cm2
  • Button front closure
  • Lapel collar
  • Left chest pocket
  • Hemmed sleeve ends
  • Side vent openings
  • Royal blue
  • Hazard rating class 1

Specifications

Product Type Flame-resistant Lab Coat
Size Large
Gender Unisex
Material Nomex IIIA, Aramid Fiber
Color Royal Blue
Closure Type Buttons
Disposable Reusable
Sterility Non-sterile
Resistance Flame
No. of Pockets 1
Collar Type Lapel
Model KNL2RBRG
Hazard Rating Class 1
Arc Flash Rating 5.6 Calories/cm2
Certifications/Compliance ASTM 1506, NFPA 2112
For Use With (Application) Chemical, Oil, Gas and Petrochemical Industries, Foundries, Cutting and Welding
Includes Side Vent Openings
Weight (English) 6 oz.
Weight (Metric) 0.17 kg
Show More Show Less