Learn More
Bulwark™ FR Nomex™ IIIA HRC1 6 oz. Uniform Shirts
Protection: Arc Rating ATPV 5.7 calories/cm2
Supplier: Bulwark™ FR SND6RBLNXL
Alert:
Not recommended for use around any molten substances, welding operations, or in a critical static control application.
Description
- Flame-resistant, 6 oz. Nomex IIIA
- Banded, topstitched collar
- Topstitched cuffs with adjustable button closures
- Placket front with button closure
- Tailored sleeve placket
Specifications
| X-Large (Long) | |
| Royal Blue | |
| 6 oz. |
| Nomex IIIA | |
| 1 |