missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ FR Men's Lightweight Nomex™ Flame Resistant Uniform Shirt

Catalog No. 191661033
Change view
Click to view available options
Color:
Navy
Royal Blue
Size:
2X-Large (Long)
2X-Large (Regular)
3X-Large (Regular)
Large (Long)
Large (Regular)
Medium (Long)
Medium (Regular)
Small (Regular)
X-Large (Long)
X-Large (Regular)
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
191661033 Royal Blue 3X-Large (Regular)
191661018 Navy Small (Regular)
191661019 Navy Medium (Regular)
191661020 Navy Large (Regular)
191661021 Navy X-Large (Regular)
191661024 Navy Medium (Long)
191661023 Navy 3X-Large (Regular)
191661026 Navy X-Large (Long)
191661027 Navy 2X-Large (Long)
191661025 Navy Large (Long)
191661022 Navy 2X-Large (Regular)
191661028 Royal Blue Small (Regular)
191661029 Royal Blue Medium (Regular)
191661031 Royal Blue X-Large (Regular)
191661030 Royal Blue Large (Regular)
191661032 Royal Blue 2X-Large (Regular)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Not recommended for use around any molten substances, welding operations, or in a critical static control application.
Catalog No. 191661033 Supplier Bulwark™ FR Supplier No. SND2RB RG 3XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Not recommended for use around any molten substances, welding operations, or in a critical static control application.

This Lightweight FR Uniform Shirt offers a tailored, professional look with CAT 1 AR/FR protection and NFPA™ 2112 compliance. Tailored sleeve placket, topstitched cuffs and straight back yoke look sharp and professional while being protective.

  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 1 Protection
  • Protection: Arc Rating ATPV 4.8 calories/cm2
  • Hazard: 1
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 4.5 oz. (150 g/m2) Nomex™ IIIA
  • Blend: 93% Nomex™ Aramid/5% Kevlar™ Aramid/2% Other Fiber
  • Closure: Button-down front and topstitched cuff with button closure
  • Collar: Banded topstitched collar
  • Pocket: Two chest pockets with button flap closures and sewn in pocket stall
  • Placket front with button closure
  • Tailored sleeve placket
  • Product Collection: Nomex™
  • Protection: ATPV Arc Rating: 4.4
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Male
  • Materials: Nomex™
  • Brand: Bulwark™

Specifications

Size 3X-Large (Regular)
Gender Men's
Material Nomex IIIA
Color Royal Blue
Closure Type Button
Hazard Risk Category 1
Arc Flash Rating ATPV 4.8 Calories/cm2
Certifications/Compliance NFPA 2112, NFPA 70E, Category 1 Protection
Weight (English) 4.5 oz.