missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ FR Men's Lightweight Nomex™ Flame Resistant Uniform Shirt
This Lightweight FR Uniform Shirt offers a tailored, professional look with CAT 1 AR/FR protection and NFPA™ 2112 compliance. Tailored sleeve placket, topstitched cuffs and straight back yoke look sharp and professional while being protective.
Supplier: Bulwark™ FR SND2RBRGS
Alert:
Not recommended for use around any molten substances, welding operations, or in a critical static control application.
Description
- Certifications Level 1: NFPA 2112, NFPA 70E
- Certifications Level 2: Category 1 Protection
- Protection: Arc Rating ATPV 4.8 calories/cm2
- Hazard: 1
- Primary Closure: Button
- Wash Care: Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame resistant, 4.5 oz. (150 g/m2) Nomex™ IIIA
- Blend: 93% Nomex™ Aramid/5% Kevlar™ Aramid/2% Other Fiber
- Closure: Button-down front and topstitched cuff with button closure
- Collar: Banded topstitched collar
- Pocket: Two chest pockets with button flap closures and sewn in pocket stall
- Placket front with button closure
- Tailored sleeve placket
- Product Collection: Nomex™
- Protection: ATPV Arc Rating: 4.4
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Regular Fit
- Gender: Male
- Materials: Nomex™
- Brand: Bulwark™
Specifications
| Small (Regular) | |
| Nomex IIIA | |
| Button | |
| ATPV 4.8 Calories/cm2 | |
| 4.5 oz. |
| Men's | |
| Royal Blue | |
| 1 | |
| NFPA 2112, NFPA 70E, Category 1 Protection |