missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ FR Men's Lightweight Nomex™ Flame Resistant Uniform Shirt

This Lightweight FR Uniform Shirt offers a tailored, professional look with CAT 1 AR/FR protection and NFPA™ 2112 compliance. Tailored sleeve placket, topstitched cuffs and straight back yoke look sharp and professional while being protective.

Supplier:  Bulwark™ FR SND2NVRG3XL

 View more versions of this product

Catalog No. 19-166-1023


Only null left
Explore available promotions

Alert:

Not recommended for use around any molten substances, welding operations, or in a critical static control application.


Description

Description

  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 1 Protection
  • Protection: Arc Rating ATPV 4.8 calories/cm2
  • Hazard: 1
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 4.5 oz. (150 g/m2) Nomex™ IIIA
  • Blend: 93% Nomex™ Aramid/5% Kevlar™ Aramid/2% Other Fiber
  • Closure: Button-down front and topstitched cuff with button closure
  • Collar: Banded topstitched collar
  • Pocket: Two chest pockets with button flap closures and sewn in pocket stall
  • Placket front with button closure
  • Tailored sleeve placket
  • Product Collection: Nomex™
  • Protection: ATPV Arc Rating: 4.4
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Male
  • Materials: Nomex™
  • Brand: Bulwark™
Specifications

Specifications

3X-Large (Regular)
Nomex IIIA
Button
ATPV 4.8 Calories/cm2
4.5 oz.
Men's
Navy
1
NFPA 2112, NFPA 70E, Category 1 Protection
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions