missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Excel FR™ Lab Coats

Catalog No. 19816950
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19816950 Medium
19816949 Small
19816952 X-Large
19816953 2X-Large
19816951 Large
19295000 3X-Large
19295001 4X-Large
19816756 X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19816950 Supplier Bulwark™ Supplier No. KEL2LB RG M

Protection: Arc Rating ATPV 7.7 calories/cm2

  • Flame-resistant, 7 oz. 100% cotton
  • Button-front closure with lapel collar
  • Hemmed sleeve ends
  • Side-vent openings
  • Light blue
  • Hazard rating class 1

Specifications

Product Type Flame-resistant Lab Coat
Size Medium
Gender Unisex
Material Cotton
Color Light Blue
Closure Type Buttons
Cuff Style Hemmed
Disposable Reusable
Sterility Non-sterile
Resistance Flame
No. of Pockets 3
Collar Type Lapel
Model KEL2LBRG
Size (English) Chest 40 to 42 in.
Size (Metric) Chest 101.6 to 106.68 cm
Hazard Rating Class 1
Arc Flash Rating 7.7 Calories/cm2
Certifications/Compliance ASTM F2302, NFPA 70E, ASTM F1506
For Use With (Application) Chemical, Oil, Gas, Petrochemical, Electrical, Utilities
Includes Side Vent Openings
Length (English) 42.5 in.
Length (Metric) 107.95 cm
Weight (English) 7 oz.
Weight (Metric) 0.19 kg
Show More Show Less