missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Excel FR™ Lab Coats

Catalog No. 19295000
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19295000 3X-Large
19816950 Medium
19816949 Small
19816952 X-Large
19816953 2X-Large
19816951 Large
19295001 4X-Large
19816756 X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19295000 Supplier Bulwark™ Supplier No. KEL2LB RG 3XL
Only null left

Protection: Arc Rating ATPV 7.7 calories/cm2

  • Flame-resistant, 7 oz. 100% cotton
  • Button-front closure with lapel collar
  • Hemmed sleeve ends
  • Side-vent openings
  • Light blue
  • Hazard rating class 1

Specifications

Product Type Flame-resistant Lab Coat
Size 3X-Large
Gender Unisex
Material Cotton
Color Light Blue
Closure Type Buttons
Disposable Reusable
Sterility Non-sterile
Resistance Flame
Collar Type Lapel
Model KEL2LBRG
Hazard Rating Class 1
Arc Flash Rating 7.7 Calories/cm2
Certifications/Compliance ASTM F2302, NFPA 70E, ASTM F1506
For Use With (Application) Chemical, Oil, Gas, Petrochemical, Electrical, Utilities
Includes Side Vent Openings
Item Description Button Front
Weight (English) 7 oz.
Weight (Metric) 0.19 kg
Show More Show Less