missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ FR Excel FR™ Flame-Resistant Work Shirt

Catalog No. 19295106
Change view
Click to view available options
Weight (English):
7 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
19295106 7 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19295106 Supplier Bulwark™ FR Supplier No. SEW2LB RG XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use in foundries, utilities and petrochemical industries

  • Flame-resistant, 7 oz. fabric
  • 100% cotton blend
  • Care: home and industrial wash
  • Two-piece, lined collar
  • One-piece lined cuffs with button closures
  • Hemmed front with button closure
  • Sleeve vent
  • Hazard Rating Class 1
  • Arc Rating: ATPV 7.7 calories/cm2

† Long Sleeve/Body adds about 2 in. to the sleeve and tail of the garment.

Specifications

Size X-Large (Regular)
Material Cotton
Color Light Blue
Arc Flash Rating ATPV 7.7 Calories/cm2
Weight (English) 7 oz.