missing translation for 'onlineSavingsMsg'
Learn More

Red Kap™ Excel FR™ Flame-Resistant Contractor Coveralls

Catalog No. 19295128
Change view
Click to view available options
Size (English):
38 in.
40 in.
42 in.
44 in.
46 in.
48 in.
50 in.
52 in.
54 in.
56 in.
58 in.
60 in.
Height (Metric):
1.55 to 1.92 m
1.73 to 1.55 m
Size (Metric) Waist:
97.7 cm
102.8 cm
107.9 cm
113 cm
118.1 cm
123.1 cm
128.2 cm
133.3 cm
138.4 cm
143.5 cm
148.5 cm
153.6 cm
Style:
Long
Regular
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English) Height (Metric) Size (Metric) Waist Style
19295128 46 in. 1.73 to 1.55 m 118.1 cm Regular
19295138 46 in. 1.55 to 1.92 m 118.1 cm Long
19295139 48 in. 1.55 to 1.92 m 123.1 cm Long
19295124 38 in. 1.73 to 1.55 m 97.7 cm Regular
19295125 40 in. 1.73 to 1.55 m 102.8 cm Regular
19295126 42 in. 1.73 to 1.55 m 107.9 cm Regular
19295130 50 in. 1.73 to 1.55 m 128.2 cm Regular
19295131 52 in. 1.73 to 1.55 m 133.3 cm Regular
19295129 48 in. 1.73 to 1.55 m 123.1 cm Regular
19295127 44 in. 1.73 to 1.55 m 113 cm Regular
19295132 54 in. 1.73 to 1.55 m 138.4 cm Regular
19295133 56 in. 1.73 to 1.55 m 143.5 cm Regular
19295134 58 in. 1.73 to 1.55 m 148.5 cm Regular
19295135 60 in. 1.73 to 1.55 m 153.6 cm Regular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 19295128 Supplier Red Kap™ Supplier No. CEC2NV RG 46
Only null left

Unbeatable protection for foundries, flame-cutting and welding

  • Flame-resistant 100% cotton twill
  • Two-way concealed Nomex™ taped brass breakaway zipper; concealed snap at top of zipper and at neck
  • One-piece, top-stitched, lay-flat collar
  • Hemmed sleeves
  • Two set-in front pockets, one chest pocket, two patch hip pockets
  • Side vent openings
  • Hazard Rating Class 2
  • Arc Rating: ATPV 11.2 calories/cm2
  • Khaki or Navy Blue

Specifications

Gender Men's
Material Cotton
Color Navy
Neck Style Collared
Closure Type 2-way Zipper
Resistance Flame
Collar Type Lay flat
Arc Flash Rating 10.6 Calories/cm2
Certifications/Compliance ASTM F2302, NFPA 70E™, ASTM F1506, NFPA 2112
No. of Pockets 5
Size (English) 46 in.
Size (English) Chest 44.5 to 46 in.
Size (English) Waist 46.5 in.
Size (Metric) Waist 118.1 cm
Height (English) 5.7 to 5.11 ft.
Height (Metric) 1.73 to 1.55 m
Style Regular
Show More Show Less