missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Red Kap™ Excel FR™ Flame-Resistant Contractor Coveralls
Unbeatable protection for foundries, flame-cutting and welding
Supplier: Red Kap™ CEC2NVRG38
Description
- Flame-resistant 100% cotton twill
- Two-way concealed Nomex™ taped brass breakaway zipper; concealed snap at top of zipper and at neck
- One-piece, top-stitched, lay-flat collar
- Hemmed sleeves
- Two set-in front pockets, one chest pocket, two patch hip pockets
- Side vent openings
- Hazard Rating Class 2
- Arc Rating: ATPV 11.2 calories/cm2
- Khaki or Navy Blue
Specifications
| Men's | |
| Navy | |
| 2-way Zipper | |
| Lay flat | |
| ASTM F2302, NFPA 70E™, ASTM F1506, NFPA 2112 | |
| 38 in. | |
| 38.5 in. | |
| 5.7 to 5.11 ft. | |
| Regular |
| Cotton | |
| Collared | |
| Flame | |
| 10.6 Calories/cm2 | |
| 5 | |
| 36.5 to 38 in. | |
| 97.7 cm | |
| 1.73 to 1.55 m |