missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ FR Men's Heavyweight Excel FR™ Comfortouch™ Flame Resistant Deluxe Insulated Bib Overall
If you're looking for the ultimate in rugged durability, warmth, and FR protection, then this Heavyweight Brown Duck Bib Overall is made for you. With CAT 4 protection, insulation throughout and side vent openings, these overalls with keep you warm and comfortable while they protect you.
$629.14 - $629.14
Specifications
| Closure Type | Hip Zipper with Concealed Snap |
|---|---|
| Color | Brown |
| Material | Cotton, Nylon |
| Construction | Cotton, Nylon, Modacrylic lining |
| For Use With (Application) | Excellent barrier to dry particles, microorganisms and non-hazardous liquids |
| Catalog Number | Mfr. No. | Size | Style | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Style | Price | Quantity | ||||
|
19-080-313
|
Bulwark™ FR
BLN4BDRGXL |
X-Large | Regular |
Each for $629.14
|
|
||||
Description
- Primary Closure: Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Shell: Flame resistant, 11 oz. (390 g/m2) Duck, Water repellent
- Blend: Shell: 88% Cotton/12% Nylon, Lining: 100% Cotton, Insulation: 100% Modacrylic
- Closure: Elasticized shoulder straps with non-conductive adjustable hardware
- Pocket: One large front bib pocket with pencil stall and two set-in front swing pockets, One watch pocket and two large hip pockets, Two-piece rule pocket and one tool pocket
- Features
- Reinforced double knees
- One large front bib pocket with pencil stall
- Two set-in front swing pockets
- Elasticized shoulder straps with non-conductive adjustable hardware
- Product Collection: Excel FR Comfortouch, Canvas
- Protection: ATPV Arc Rating: 44
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Designed to be worn over other clothing. Fit over work boots.
- Gender: Male
- Materials: Cotton/Nylon
- Brand: Bulwark™
Specifications
| Hip Zipper with Concealed Snap | |
| Cotton, Nylon | |
| Excellent barrier to dry particles, microorganisms and non-hazardous liquids |
| Brown | |
| Cotton, Nylon, Modacrylic lining | |
| 8 |