missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ FR Men's Midweight Excel FR™ Comfortouch™ Flame Resistant Deluxe Insulated Bib Overall

Catalog No. 19080319
Change view
Click to view available options
Size:
Small
Large
Style:
Regular
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Style
19080319 Small Regular
19080321 Large Regular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19080319 Supplier Bulwark™ FR Supplier No. BLC8NV RG S

For cold jobs, this Midweight Insulated Bib Overall offers unbeatable warmth with CAT 3 AR/FR protection and deluxe features like rugged, double-reinforced knees. Large patch pockets and bonus utility pockets ensure there's a place for all of your tools.

  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 3 Protection
  • Hazard:
  • Primary Closure: Concealed Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Shell: Flame resistant, 7 oz. (235 g/m2) Twill, Water-repellent, Insulation Package: 14 oz. (474 g/m2)
  • Blend: Shell: 88% Cotton/12% Nylon, Insulation Package: 50% Modacrylic/50% Cotton
  • Closure: Elasticized shoulder straps with non-conductive adjustable hardwear
  • Pocket: Five large patch pockets and hip pocket with closure
  • Features:
    • Reinforced double knees
    • Elasticized shoulder straps with non-conductive adjustable hardwear
  • Product Collection: Excel FR Comfortouch
  • Protection: ATPV Arc Rating: 36
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Designed to be worn over other clothing. Fit over work boots.
  • Gender: Male
  • Materials: Cotton/Nylon
  • Brand: Bulwark™

Specifications

Closure Type Hip Zipper with Concealed Snap
Color Navy
Material Cotton, Nylon
Size Small
Construction Cotton, Nylon, Modacrylic lining
For Use With (Application) Excellent barrier to dry particles, microorganisms and non-hazardous liquids
Length (English) Inseam 31.5 in.
Length (Metric) Inseam 80 cm
No. of Pockets 5
Size (English) Chest 32.5 to 34.5 in.
Size (English) Waist 32.5 to 34.5 in.
Size (Metric) Chest 82.5 to 87.6 cm
Size (Metric) Waist 82.5 to 87.6 cm
Style Regular
Waist Style Elastic
Show More Show Less