Learn More
Bullard™ CC20 Series of DuPont™ Tychem™ 2000 Hoods
For use with select Bullard™ supplied air or powered air respirators.
$109.03 - $109.03
Specifications
| Assigned Protection Factor (APF) | 1000 |
|---|---|
| Material | Tychem QC |
| Product Type | Double Bib Hood |
| For Use With (Application) | Pharmaceutical, Chemical, Pesticides, Spray Painting and Coating, Nuclear |
| For Use With (Equipment) | Supplied Air Respirator (CC20 Series) or Powered Air Respirator (EVA, EVAHL, PA20, PA30 or PA30IS Series) |
| Catalog Number | Mfr. No. | Suspension Type | Connection Type | Lens Type | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Suspension Type | Connection Type | Lens Type | Price | Quantity | |||
|
19-700-100
|
Bullard™
20TICT |
Ratchet Suspension | Threaded | PETG |
Each for $109.03
Case of 10 Each for $1,024.20
|
|
|||
Description
- Loose fitting respirator double bib hoods
- Choose between ratchet or no suspension models
- Threaded connection for PAPR
- DuPont Tychem 2000 material construction
- 1,000 Assigned Protection Factor (APF)
Specifications
| 1000 | |
| Double Bib Hood | |
| Supplied Air Respirator (CC20 Series) or Powered Air Respirator (EVA, EVAHL, PA20, PA30 or PA30IS Series) |
| Tychem QC | |
| Pharmaceutical, Chemical, Pesticides, Spray Painting and Coating, Nuclear |