missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bullard™ RT Series Supplied-Air Respirator Breathing Tubes
Description
- Component for use with RT series respirators
- Choose between heavy-duty or lightweight options
- Breathing tubes must be used in conjunction with hood, flow control device, and air hose
Specifications
Specifications
| Description | Lightweight, disposable breathing tube |
| Color | Gray |
| Product Type | Breathing Tube |
| For Use With (Equipment) | RT Series supplied-air respirators |
| Includes | S18051 hose clamp |
| Model | RTBT |