Learn More
Bullard™ Supplied-Air Respirator Accessories, Belts
Catalog No. 361003806
Change view
Click to view available options
Material:
Nylon
Vinyl Decon
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
361003806 Vinyl Decon
19059266 Nylon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361003806 Supplier Bullard™ Supplier No. 36501
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Replacement belt features durable nylon or vinyl construction for use with belt-mounted SAR climate control devices.
Specifications
For Use With (Equipment) 20LF Series Airline Respirators
Material Vinyl Decon
Model 36501
Overall Length (English) 54 in.
Overall Length (Metric) 137 cm
Product Type Belt