Learn More
Bullard™ Standard Series S62 Vented Hardhats
Low-profile shell design with four-point suspension system
Supplier: Bullard™ 62PBP
Description
Setting the Standard. Sleek, low-profile design with a patented Flex-Gear ratchet suspension option.
Features
- Absorbent, replaceable cotton brow pad is pillowed for maximum comfort
- Ideal for construction and everyday work environments
- One of the lightest helmets of its class
- Vented, providing increased ventilation and comfort
- Accessory slots allow attachment of accessories such as earmuffs and face protection
- Vertical height adjustment capability (2 front, 2 rear)
- Choose 4-point pinlock suspension or 4-point Flex-Gear ratchet suspension
- Pinlock suspension provides incremental sizing of 1/16 hat size (5mm), for a secure fit
- Ratchet suspension can be adjusted between hat sizes 6 1/2-8
Order Info
Snap-on replacement brow pads are available (see below).
Compliance
ANSI Z89.1-2003, Type I, Class E and G (S61 Model); Class C (S62 Vented).
Specifications
| ANSI Z89.1, Type I, Class C | |
| Pacific Blue | |
| 4-Point Pin-Lock |