Learn More
Bullard™ Standard Series S62 Vented Hardhats
Low-profile shell design with four-point suspension system
Supplier:  Bullard™ 62PBR
 View more versions of this product
Catalog No. 19-160-454
Only null left
Explore available promotions
Description
Description
Setting the Standard. Sleek, low-profile design with a patented Flex-Gear ratchet suspension option.
Features
  • Absorbent, replaceable cotton brow pad is pillowed for maximum comfort
  • Ideal for construction and everyday work environments
  • One of the lightest helmets of its class
  • Vented, providing increased ventilation and comfort
  • Accessory slots allow attachment of accessories such as earmuffs and face protection
  • Vertical height adjustment capability (2 front, 2 rear)
Pinlock or Ratchet Suspension
  • Choose 4-point pinlock suspension or 4-point Flex-Gear ratchet suspension
  • Pinlock suspension provides incremental sizing of 1/16 hat size (5mm), for a secure fit
  • Ratchet suspension can be adjusted between hat sizes 6 1/2-8
Order Info
Snap-on replacement brow pads are available (see below).
Compliance
ANSI Z89.1-2003, Type I, Class E and G (S61 Model); Class C (S62 Vented).
Specifications
Specifications
ANSI Z89.1, Type I, Class C
Pacific Blue
Flex-Gear Ratchet
Product Suggestions
Product Suggestions