Learn More
Bullard™ Standard Series S62 Vented Hardhats
Catalog No. 19160444
Change view
Click to view available options
Color:
Black
Blue
Charcoal Brown
Dove Gray
Forest Green
Hi-vis Orange
Hi-vis Yellow
Kelly Green
Kentucky Blue
Orange
Pacific Blue
Red
Tan
White
Yellow
Suspension Type:
4-Point Flex-Gear Ratchet
4-Point Pin-Lock
Flex-Gear Ratchet
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19160444 Red 4-Point Flex-Gear Ratchet
19160443 White 4-Point Flex-Gear Ratchet
19160447 Kentucky Blue 4-Point Flex-Gear Ratchet
19160448 Yellow 4-Point Flex-Gear Ratchet
19160449 Charcoal Brown Flex-Gear Ratchet
19160452 Kelly Green Flex-Gear Ratchet
19160450 Orange 4-Point Flex-Gear Ratchet
19160455 Tan 4-Point Flex-Gear Ratchet
19160453 Dove Gray Flex-Gear Ratchet
19160456 Black Flex-Gear Ratchet
19160458 Red 4-Point Pin-Lock
19160459 Orange 4-Point Pin-Lock
19160461 Blue 4-Point Pin-Lock
19160460 Forest Green 4-Point Pin-Lock
19160465 Hi-vis Yellow 4-Point Pin-Lock
19160464 Hi-vis Orange 4-Point Pin-Lock
19160463 Charcoal Brown 4-Point Pin-Lock
19160468 Pacific Blue 4-Point Pin-Lock
19160466 Kelly Green 4-Point Pin-Lock
19160467 Dove Gray 4-Point Pin-Lock
19160469 Tan 4-Point Pin-Lock
19160470 Black 4-Point Pin-Lock
19160454 Pacific Blue Flex-Gear Ratchet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Catalog No. 19160444 Supplier Bullard™ Supplier No. 62RDR
Low-profile shell design with four-point suspension system
Setting the Standard. Sleek, low-profile design with a patented Flex-Gear ratchet suspension option.
Features
  • Absorbent, replaceable cotton brow pad is pillowed for maximum comfort
  • Ideal for construction and everyday work environments
  • One of the lightest helmets of its class
  • Vented, providing increased ventilation and comfort
  • Accessory slots allow attachment of accessories such as earmuffs and face protection
  • Vertical height adjustment capability (2 front, 2 rear)
Pinlock or Ratchet Suspension
  • Choose 4-point pinlock suspension or 4-point Flex-Gear ratchet suspension
  • Pinlock suspension provides incremental sizing of 1/16 hat size (5mm), for a secure fit
  • Ratchet suspension can be adjusted between hat sizes 6 1/2-8
Order Info
Snap-on replacement brow pads are available (see below).
Compliance
ANSI Z89.1-2003, Type I, Class E and G (S61 Model); Class C (S62 Vented).
Specifications
Certifications/Compliance Meets ANSI Z89.1-2003; Type I; Class E and G (S61 Model); Class C (S62 Vented).
Color Red
Size Standard
Suspension Type 4-Point Flex-Gear Ratchet