Learn More
Bullard™ Standard Series S61 Unvented Hardhats
Low-profile shell design with four-point suspension system
Supplier: Bullard™ 61CBR
Description
Setting “The Standard”. Sleek, low-profile design with a patented “Flex-Gear” Ratchet suspension option.
Features
- Absorbent, replaceable cotton brow pad is pillowed for maximum comfort
- Ideal for construction and everyday work environments
- One of the lightest helmets of its class
- Unvented; see Model S62 for vented style
- Accessory slots allow attachment of accessories such as earmuffs and face protection
- Vertical height adjustment capability (2 front, 2 rear)
- Choose 4-point pinlock suspension or 4-point “Flex-Gear” ratchet suspension
- Pinlock suspension provides incremental sizing of 1/16 hat size (5mm), for a secure fit
- Ratchet suspension can be adjusted between hat sizes 61/2-8
Order Info
Snap-on replacement brow pads are available (see below).
Compliance
ANSI Z89.1-2003, Type I, Class E and G (S61 Model); Class C (S62 Vented).
Specifications
| ANSI/ISEA Z89.1 Type I, Class E & G | |
| Charcoal Brown | |
| Flex-Gear Ratchet |