Learn More
Bullard™ Standard Series S61 Unvented Hardhats
Catalog No. 19160415
Change view
Click to view available options
Color:
Black
Charcoal Brown
Dove Gray
Forest Green
Hi-vis Orange
Hi-vis Yellow
Kelly Green
Kentucky Blue
Orange
Pacific Blue
Red
Tan
White
Yellow
Suspension Type:
4-Point Flex-Gear Ratchet
4-Point Pin-Lock
Flex-Gear Ratchet
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19160415 White 4-Point Pin-Lock
19160417 Orange 4-Point Pin-Lock
19160418 Forest Green 4-Point Pin-Lock
19160416 Red 4-Point Pin-Lock
19160420 Yellow 4-Point Pin-Lock
19160425 Dove Gray 4-Point Pin-Lock
19160422 Hi-vis Orange 4-Point Pin-Lock
19160423 Hi-vis Yellow 4-Point Pin-Lock
19160421 Charcoal Brown 4-Point Pin-Lock
19160427 Tan 4-Point Pin-Lock
19160426 Pacific Blue 4-Point Pin-Lock
19160428 Black 4-Point Pin-Lock
19160429 White 4-Point Flex-Gear Ratchet
19160433 Kentucky Blue 4-Point Flex-Gear Ratchet
19160436 Hi-vis Orange 4-Point Flex-Gear Ratchet
19160438 Kelly Green Flex-Gear Ratchet
19160432 Forest Green Flex-Gear Ratchet
19160439 Dove Gray Flex-Gear Ratchet
19160435 Charcoal Brown Flex-Gear Ratchet
19160440 Pacific Blue Flex-Gear Ratchet
19160441 Tan Flex-Gear Ratchet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 19160415 Supplier Bullard™ Supplier No. 61WHP
Only null left
Low-profile shell design with four-point suspension system
Setting “The Standard”. Sleek, low-profile design with a patented “Flex-Gear” Ratchet suspension option.
Features
  • Absorbent, replaceable cotton brow pad is pillowed for maximum comfort
  • Ideal for construction and everyday work environments
  • One of the lightest helmets of its class
  • Unvented; see Model S62 for vented style
  • Accessory slots allow attachment of accessories such as earmuffs and face protection
  • Vertical height adjustment capability (2 front, 2 rear)
Pinlock or Ratchet Suspension
  • Choose 4-point pinlock suspension or 4-point “Flex-Gear” ratchet suspension
  • Pinlock suspension provides incremental sizing of 1/16 hat size (5mm), for a secure fit
  • Ratchet suspension can be adjusted between hat sizes 61/2-8
Order Info
Snap-on replacement brow pads are available (see below).
Compliance
ANSI Z89.1-2003, Type I, Class E and G (S61 Model); Class C (S62 Vented).
Specifications
Certifications/Compliance ANSI/ISEA Z89.1 Type I, Class E & G
Color White
Suspension Type 4-Point Pin-Lock