Learn More
Bullard™ Standard Series S51 Caps
Catalog No. 173591B
Change view
Click to view available options
Color:
Black
Charcoal Brown
Forest Green
Hi-vis Orange
Orange
Pacific Blue
Red
Tan
White
Yellow
Suspension Type:
4-Point Pin-Lock
4-Point Ratchet
Flex-Gear Ratchet
Standard
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
173591B Yellow Flex-Gear Ratchet
173591C Red Flex-Gear Ratchet
17986226 Orange Standard
19098025 Hi-vis Orange 4-Point Pin-Lock
19098028 Hi-vis Orange 4-Point Ratchet
19160475 Tan 4-Point Pin-Lock
19160477 Charcoal Brown Flex-Gear Ratchet
19160474 Pacific Blue 4-Point Pin-Lock
19160479 Pacific Blue Flex-Gear Ratchet
19160476 Black 4-Point Pin-Lock
19160481 Black Flex-Gear Ratchet
173591A White Flex-Gear Ratchet
19082860 Forest Green Flex-Gear Ratchet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 missing translation for 'options'
Catalog No. 173591B missing translation for 'mfr' Bullard™ missing translation for 'supplierNo' 51 YLR
Dielectric hard cap is one of the lightest safety caps available
  • Constructed of polyethylene
  • Soft poly browpad absorbs moisture
  • Rain trough provides extra protection from snow, rain, dust and dirt
  • Accessory slots accommodate a variety of optional attachments
  • Available with standard four-point suspension or four-point Flex-Gear™ ratchet suspension featuring two vertical adjustments
  • Wide 0.75 in. (1.9cm) crown straps and low nape strap keep caps securely in place
Order Info
Note: There is a minimum order quantity of 20.
Compliance
ANSI/ISEA Z89.1-2009, Type I, Class E & G and CSA Z94.1-2005, Type 1, Class E & G requirements.
Specifications
Certifications/Compliance Meets ANSI Z89.1-1997 Type I; Class G and E.
Color Yellow
Material Polyethylene
Suspension Type Flex-Gear Ratchet