Learn More
Bullard™ Spectrum™ Series Full-Facepiece Airline Respirators
Continuous-flow respirators feature lightweight facepieces with a wide-angle lens for a clear field of view.
$944.29 - $944.29
Specifications
| Certifications/Compliance | NIOSH |
|---|---|
| Includes | Headstrap, breathing tube assembly with flow control device and nylon belt |
| Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Size | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Size | Price | Quantity | |||
|
29-663-318
|
Bullard™
SPEC40L |
High Pressure Respirator Assembly | Breathing air compressor | Medium/Large |
Each for $944.29
|
|
|||
Description
- Continuous-flow, high-pressure model includes breathing tube and flow valve for compressor use
- Continuous-flow, low-pressure model includes breathing tube and constant flow valve for air pump use
- Silicone flange seals for comfortable fit
- SnapFit™ design for easy donning
- Clear silicone nosecup and low-profile regulator minimize interference with vision and other PPE
- Regulator allows donning without free flow of air, conserving air supply
- Swivel connector provides free movement of regulator hose for comfort and maneuverability
- All models equipped with medium/large facepiece
- Can be used with V10 and V20 Series breathing air hoses (order separately)
Compliance
NIOSH approved.
Specifications
| NIOSH | |
| Headstrap, breathing tube assembly with flow control device and nylon belt |