missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ Spectrum™ Replacement Facepieces

Catalog No. p-3775629
Change view
Click to view available options
Format:
Medium/Large
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
19321653 Medium/Large
19321654 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19321653 Supplier Bullard™ Supplier No. PASPECML
Only null left

For use with PA40 series powered air purifying respirators

  • Lens made of polycarbonate
  • With silicone suspension, exhalation valve, headstraps, speaker diaphragm, nose cup and nose clip
  • Optically correct
  • Convertible to SAR
  • Available in different sizes

Specifications

Product Type Full Facepiece
Format Medium/Large
For Use With (Equipment) Pa40 series powered air purifying respirator
Includes Silicone suspension, exhalation valve, headstraps, speaker diaphragm, nose cup and nose clip